• USA Home
  • Z255947 - Nalgene® 180 vacuum tubing
EMAIL THIS PAGE TO A FRIEND
Z255947 Aldrich
Nalgene® 180 vacuum tubing
I.D. × O.D. × wall 1/2 in. × 1 1/8 in. × 5/16 in.
Purchase
Properties
Related Categories Filtration Flasks, Filtration Supplies, Labware, Manufacturer Browser, Nalgene,
packaging   pkg of 10 feet
  pkg of 50 feet
mfr. no.   Nalgene
limit   28 psi max. pressure (25 °C)
  56 psi max. pressure (25 °C)
I.D. × O.D. × wall   1/2 in. × 1 1/8 in. × 5/16 in.
Description
General description
Transparent, nontoxic, autoclavable plastic tubing has thick walls for high vacuum.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?