• USA Home
  • Z255963 - Digital display stopwatch
EMAIL THIS PAGE TO A FRIEND
Z255963 Aldrich
Digital display stopwatch
Purchase
Properties
Related Categories Labware, Timers More...
Description
General description
Times up to 24h in 0.01s intervals. Split interval function and a 12/24h alarm clock, calendar, and hourly chime option.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z255971 8-Memory stopwatch, two simultaneous displays
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?