EMAIL THIS PAGE TO A FRIEND
Z255971 Aldrich
8-Memory stopwatch
two simultaneous displays
Purchase
Properties
Related Categories Labware, Timers More...
Description
General description
Times 10h (up or down) in 0.01-sec intervals. Stores up to 8 times in memory to handle multiple processes. Two simultaneous 0.01s digital displays. Shock resistant.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?