• USA Home
  • Z256099 - Dreiding stereomodels
EMAIL THIS PAGE TO A FRIEND
Z256099 Aldrich
Dreiding stereomodels
Peptide A Set, 82 parts
Purchase
Properties
Related Categories Dreiding models, Labware, Molecular Models, Molecular Models and Educational Aids
mfr. no.   Büchi
availability   not available in EU
Description
Application
Highly accurate stainless steel molecular models for demonstrating stereochemical factors in conformational analysis and in the study of reaction mechanisms.
Features and Benefits
Model units consist of push-together rods and tubes radiating from a point that represents the atomic nucleus. Bonds rotate freely to show conformational changes. Elements, angle strain, and bond types are identified by color-coded marks on the rods and tubes.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?