EMAIL THIS PAGE TO A FRIEND
Z256323 Aldrich
Wrap-it tie set
180 pieces in a PP case
Purchase
Properties
Related Categories Labware, Septa, Septa Accessories More...
availability   not available in EU
Description
General description
Contains 180 pieces in a PP case. Includes sizes from 3 1/4 to 7 1/2 in. Also includes 3 1/2 in. hanking and marker ties.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Wrap-It Tie Installation [VIDEO]
The self-locking, one-piece nylon construction of the Wrap-it tie provides an excellent way to secure rubber septa to bottles and flask joints. By using the Wrap-it tie and the installing tool, easy ...
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?