• USA Home
  • Z256692 - Kontes® 14/10 MICROFLEX® glassware kits
EMAIL THIS PAGE TO A FRIEND
Z256692 Aldrich
Kontes® 14/10 MICROFLEX® glassware kits
Basic kit
Purchase
Description
General description
Components in Basic kit refer to letters in the corresponding kit drawing.
Glassware has 14/10 joints with threaded O-ring cap seals and blue nylon connectors that require minimal clamping and support. Kits contain glassware needed to perform experiments described in the textbooks: Microscale Organic Laboratory and Organic Laboratory Techniques: A Microscale Approach
Standard Kit Components, Qty
A
Air condenser (1)
B 15 mL Centrifuge tube (1)
C Claisen adapter (1)
D Compression cap 20-400 (12)
E Connecting adapter (1)
F Jacketed condenser (1)
G 2 to 3 mL Craig recrystallization tube (1)
H Glass Craig recrystallization plug (1)
I Drying tube (1)
J 5 mL Pear-shaped flask (1)
K 10 mL Round-bottom flask (1)
L Gas delivery tube 20-400 (1)
M GC collection tube (1)
N Hickman distillation head (1)
O 0.1 mL Micro vial (1)
O 3 mL Micro vial (1)
O 5 mL Micro vial (1)
P O-ring, EP, size 112 (8)
Q Septum, PTFE/silicone (4)
R Spin vane (1)
S PE syringe (1)
T Thermometer adapter (1)
U Foam insert set (1)
U Ekono-case (1)
Basic Kit Components, Qty
C
Claisen adapter (1)
D Compression cap 20-400 (7)
F Jacketed condenser (1)
G 2 to 3 mL Craig recrystallization tube (1)
H PTFE Craig recrystallization plug (1)
I Drying tube (1)
K 10 mL Round-bottom flask (1)
N Hickman distillation head (1)
O 3 mL Micro vial (1)
O 5 mL Micro vial (1)
P O-ring, EP, size 112 (5)
Q Septum, PTFE/silicone (2)
T Thermometer adapter, (1)
U Universal inlet/vacuum adapter (1)
Foam insert set (1)
Ekono-case (1)
Legal Information
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
MICROFLEX is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z558575 Aldrich® microscale glassware kit, Economy
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?