• USA Home
  • Z256846 - Ace Liebig condenser with Ace-Thred hose connections
EMAIL THIS PAGE TO A FRIEND
Z256846 Aldrich
Ace Liebig condenser with Ace-Thred hose connections
jacket L 200 mm
Purchase
Properties
Related Categories Ace, Ace Condensers, Glass Condensers, Glassware, Labware,
Joint   Joint ST/NS24/40
mfr. no.   Ace Glass 5997132
jacket L   200 mm
Description
General description
"No Hold Up" condenser with #7 Ace-Thred hose connections for 1/4 in .i.d. tubing. Features threaded inlet/outlet connections that incorporate UHDPE connectors and nylon bushings to make easy connect/disconnect of coolant tubing. The bushing compresses the connector and a silicone O-ring for a leaktight seal.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?