• USA Home
  • Z256927 - Ace dry ice condenser-trap
EMAIL THIS PAGE TO A FRIEND
Z256927 Aldrich
Ace dry ice condenser-trap
With UHDPE tubing connections
Purchase
Description
General description
1,000 mL reservoir, 3 mm bore PTFE stopcock at bottom for draining. 9.5 mm inlet and outlet connections.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?