• USA Home
  • Z257451 - PTFE-coated deep-immersion thermometer
EMAIL THIS PAGE TO A FRIEND
Z257451 Aldrich
PTFE-coated deep-immersion thermometer
-10-360 °C temperature, immersion level, 12 in.
Purchase
Properties
Related Categories HB Instruments, Labware, Manufacturer Browser, PTFE-coated deep-immersion thermometer, Temperature,
feature   immersion level 12 in.
mfr. no.   HB Instruments
measuring range   -10-360 °C temperature
Description
General description
Engraved 1° divisions on yellow backed glass. Mercury fill with safety PTFE coating.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?