• USA Home
  • Z257869 - TempTrace temperature recorder
EMAIL THIS PAGE TO A FRIEND
Z257869 Aldrich
TempTrace temperature recorder
recording time 24 hr, white, parameter -20-120 °F temp. range, with digital temperature display
Purchase
Properties
Related Categories Electronic Meters, Labware, Temperature More...
parameter   -20-120 °F temp. range
recording time   24 hr
L × W × D   7 in. × 5 1/2 in. × 2.7 in.
color   white
Description
General description
Totally self-contained and easy-to-use. Features a corrosion resistant, high impact plastic case. Unit complete with AA battery, chart paper, pens, and manual.
Specifications:
• Temp. sensor: spiral wound, bimetallic strip transducer
• Pen accuracy: ±2% full scale
• Average response time: 11min to move 63% of scale
White comes with digital temperature display
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?