• USA Home
  • Z258016 - Temptrace II remote sensing temperature recorders
EMAIL THIS PAGE TO A FRIEND
Z258016 Aldrich
Temptrace II remote sensing temperature recorders
recording time 7 day, parameter 0-250 °C temp. range
Purchase
Properties
parameter   0-250 °C temp. range
recording time   7 day
Description
General description
With remote sensing probe to document temperatures too extreme for direct sensing recorders. Ideal for oven, incubator and freezer applications.
Supplied complete with AA battery, chart paper, pens and manual.
Specifications:
• Temp sensor: Spiral wound Bourdon tube trasducer
• Temp. Accuracy: ± 2% full scale
• Probe: external 4 in. SS with 6ft lead
• Average response time: 30 secs to more 63% scale
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?