• USA Home
  • Z258342 - Periodic table of the elements
EMAIL THIS PAGE TO A FRIEND
Z258342 Aldrich
Periodic table of the elements
Wallet size
Purchase
Properties
Related Categories Educational Aids, Labware, Molecular Models and Educational Aids More...
packaging   pkg of 5 ea
W × L   2 1/4 in. × 3 1/2 in.
Description
General description
Laminated. Excellent reference tools for students and researchers.
Poster:
Full-color. Chart information includes: atomic weight, melting point, boiling point, density, electronegativity, first ionization potential, group classifications, atomic number, oxidation states, element symbol, electronic configuration, and name. Plus special tables for Elementary Subatomic Particles and % Ionic Character of a Single Chemical Bond.
Binder:
Two-sided chart features more than 20 properties of the elements, from electron configuration and electronegativity to covalent radius and thermal conductivity.
Wallet:
Two-sided chart features properties of the elements in table format with useful chemical constants on the back.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?