• USA Home
  • Z258628 - Whatman® Schleicher & Schuell® chromatography papers
EMAIL THIS PAGE TO A FRIEND
Z258628 Aldrich
Whatman® Schleicher & Schuell® chromatography papers
Type 593, size 8 in. × 10 in.
Purchase
Properties
size   8 in. × 10 in.
Description
General description
Chromatography and blotting paper. Relatively thick paper (0.36 mm) with medium flow rate. Smooth white surface
with high wet strength. Widely used for chromatography and electrophoresis. Available in sheets and rolls.
Legal Information
Schleicher & Schuell is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of GE Healthcare
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?