• USA Home
  • Z258776 - Ace heavy-wall filtering flask
EMAIL THIS PAGE TO A FRIEND
Z258776 Aldrich
Ace heavy-wall filtering flask
capacity 1,000 mL
Synonym: filter flasks, filtering flask, filtering flasks
Purchase
Properties
Related Categories Ace, Ace Flasks, Ace Heavy-Wall Filtering Flasks, Filtering Flasks, Flasks,
material   borosilicate glass
  narrow-mouth
stopper size size   8
sterility   non-sterile
feature   closure type Stopper
packaging   pack of 1
mfr. no.   Ace Glass 698336
capacity   1,000 mL
Description
General description
With tooled neck for uniform stopper fit. Side hose connection is #11 Ace-Thred with an ultra-high-density PE tubing connector, silicone O-ring, and nylon bushing to safely connect/disconnect 3/8 in. I.D. tubing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?