• USA Home
  • Z259438 - ACE temperature controller
EMAIL THIS PAGE TO A FRIEND
Z259438 Aldrich
ACE temperature controller
Multiple output controller
Purchase
Properties
Related Categories Ace, Ace Heating, Heating, Heating Controllers, Labware,
AC input   120 V
mfr. no.   Ace Glass 1210335
H × W × D   15.9 cm × 14 cm × 20.1 cm
  6 1/4 in. × 5 1/2 in. × 7 9/32 in.
weight   5.9 kg (14 lb)overall
output   40 V
Description
General description
20, 40, 120 V AC. Recommended for use with Instatherm oil baths and other heating equipment.
Time proportional; analog set, dial in desired temperature. Controllers include an RTD sensor - 318 mm × 6 mm O.D. The ACE temperature controller does not have CSA approval.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?