• USA Home
  • Z259446 - ACE temperature controller
EMAIL THIS PAGE TO A FRIEND
Z259446 Aldrich
ACE temperature controller
Single output controller
Purchase
Properties
Related Categories Ace, Ace Heating, Heating, Heating Controllers, Labware,
AC input   120 V
mfr. no.   Ace Glass 1210232
H × W × D   15.9 cm × 14 cm × 20.1 cm
  6 1/4 in. × 5 1/2 in. × 7 9/32 in.
weight   1.8 kg (5 lb)overall
Description
General description
Time proportional; analog set, dial in desired temperature. Controllers include an RTD sensor - 318 mm × 6 mm O.D. The ACE temperature controller does not have CSA approval.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?