EMAIL THIS PAGE TO A FRIEND
Z259713 Aldrich
Purge assemblies
tee-purge, stainless steel, inlet 1/4 in. male NPT
Purchase
Properties
Related Categories Aldrich, Aldrich Gas Equipment, Aldrich Valves (Gas), Gas Equipment Product Guide, Gas Lecture Bottle Accessories,
material   stainless steel
feature   tee-purge
fitting   inlet 1/4 in. male NPT
Description
General description
Purge assemblies remove vapor contaminants and moisture, reducing the chance of reaction with the process gas which could damage the regulator and equipment. When used properly, a purge assembly can also enhance operator safety. Max. operating pressure is 3,000 psig. Please request Technical Information Bulletin No. AL-202 for cross-purge techniques.
Tee-purge: Two -valve system allows purge gas to remove atmospheric contamination prior to start-up or after cylinder change.
Cross-purge: Same functions as the tee-purge, with the option of vacuum for a more effective purge. The three-valve system also features an inlet isolation valve and a vent valve.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?