• USA Home
  • Z259802 - Igloo® thermoelectric warmer/cooler
EMAIL THIS PAGE TO A FRIEND
Z259802 Aldrich
Igloo® thermoelectric warmer/cooler
AC/DC converter, AC input 220 V AC
Purchase
Description
General description
• Cools to 40 °F below ambient temperature or warms to 140 °F
• Thermoelectric solid-state heat pump
• Lightweight Ultratherm® insulation
• Plugs into cigarette lighter in cars and trucks
• Converter available for 120 and 240 V AC operation
Powers Igloo thermoelectrics using AC current.
Legal Information
Igloo is a registered trademark of Igloo Products Corp.
Ultratherm is a registered trademark of Igloo Products Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?