• USA Home
  • Z259985 - FORTUNA® Optifix® Solvent bottle-top dispenser
EMAIL THIS PAGE TO A FRIEND
Z259985 Aldrich
FORTUNA® Optifix® Solvent bottle-top dispenser
Solvents model, 0.2-1.0 mL volume
Purchase
Description
General description
For dispensing solvents
Features:
• Dispensing piston made of glass
• Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
• FIX adapter made of white PP up to 10 mL; FIX adapter made of solid white PP from 30 mL size
• Valve block made of PTFE, no metal springs in the valve - no corrosion
• Ergonomic finger protection cap
• Fast and easy volume setting with additional fine adjustment
• Optifix Solvent dispenser available up to 500 mL
Supplied as Optifix Solvent dispenser, 2 PP adapters GL40 + GL45 & 1 PTFE intake tube
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?