• USA Home
  • Z260797 - Igloo® all-plastic cooler
EMAIL THIS PAGE TO A FRIEND
Z260797 Aldrich
Igloo® all-plastic cooler
capacity 2 gal (7.6 L)
Purchase
Properties
Related Categories Cryoware, Ice and Insulated Containers, Igloo Containers, Labware
H × I.D.   13 1/4 in. × 8 1/8 in.
capacity   2 gal (7.6 L)
Description
General description
Industrial grade, HDPE yellow body, red lid, molded-in handles, Ultratherm® insulation, and recessed plastic spigots. Suitable for temporary storage of ice water, dry ice, or for use as hot/cold bath. White FDA grade liner resists stains and odors. 2 and 3 gal sizes have plastic bail handle. 5 and 10 gal coolers have plastic side handles and nylon cords to secure lid to side handle.
Legal Information
Igloo is a registered trademark of Igloo Products Corp.
Ultratherm is a registered trademark of Igloo Products Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?