• USA Home
  • Z261033 - Nalgene® polypropylene separatory funnel
EMAIL THIS PAGE TO A FRIEND
Z261033 Aldrich
Nalgene® polypropylene separatory funnel
capacity 500 mL
  •  eCl@ss 32035600
Purchase
Properties
Related Categories Funnels, Labware, Manufacturer Browser, Nalgene, Nalgene Funnels,
mfr. no.   Nalgene® 4300
capacity   500 mL
closure size   43 mm
Description
General description
Break-resistant and nearly transparent. Phase interface visible down to stopcock. PTFE stopcock and Tefzel® screw closure are leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?