• USA Home
  • Z261750 - Stereoselective Synthesis
EMAIL THIS PAGE TO A FRIEND
Z261750 Aldrich
Stereoselective Synthesis
  •  ISBN-10 0471952508
  •  ISBN-13 978-047195250-3
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   R. Atkinson, John Wiley & Sons, Ltd, 1995, 542 pp., hard cover
mfr. no.   Wiley
Description
General description
Introduces a new, simplified classification for stereoselective reactions based on changes in the number of chiral centres taking place, emphasizing the three-dimensional representations of transition states in which stereoselectivity is determined. Using stereochemical vocabulary in accordance with IUPAC rules, it covers analysis of factors in which asymmetric induction occurs, the interplay between steric and electronic effects, the importance of conformational control, advantages of intramolecularity and the majority of important reaction types used in modern stereoselective synthesis.
Table of Contents
PARTIAL TABLE OF CONTENTS:
Type 0 Reactions: Reactions in Which No New Chiral Centres Are Created.
Type I Reactions: Those Proceeding with Either Inversion or with Retention of Configuration at a Single Chiral Center.
Simple Chirality Transfer Reactions: Those in Which a Single Chiral Center Is Transferred with Concomitant Migration of One or More Double Bonds.
Type II Reactions: Simple Diastereoselectivity in 1,2-Additions to Alkenes, Diels-Alder and 1,3-Dipolar Cycloadditions and the ene Reaction.
Type II Reactions: Occasional Diastereoselectivity.
Type III Reactions: Those Involving Asymmetric Induction.
Type III Reactions of Diastereotopic and Enantiotopic Atoms and Groups; Enzyme Catalysed Reactions.
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?