EMAIL THIS PAGE TO A FRIEND
Z261831 Aldrich
Flash arrestor
Brass body
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Regulators, Labware More...
description   with butyl rubber seat
mfr. no.   Matheson 6103
measuring range   ≤15 psig operating pressure, (Acetylene)
  ≤15 psig operating pressure, (Natural gas)
  ≤150 psig operating pressure, (Oxygen)
  ≤50 psig operating pressure, (Hydrogen)
  ≤50 psig operating pressure, (Propane)
Description
General description
Install in-line for a flammable gas. In the event of a flashback, the flash arrestor diverts and extinguishes the flame. The arrestor′s shut-off valve closes, eliminating continued gas feed. Reusable 1/4 in. NPTF connections. The 316 SS flash arrestor is recommended for protecting hydrogen systems in high-purity applications.
Other Notes
Factory Mutual Approved Services only.
Warning
For HYDROGEN Service
These arrestors should not be used in installations where it is possible for pure oxygen or gas mixtures containing greater than 20% oxygen to enter the hydrogen system. Models Z261831 and Z261858 flash arrestors provide excellent flashback protection for situations in which air accidentally enters the hydrogen system and a flashback results.
Not for use with SILANE or NITROUS OXIDE
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?