• USA Home
  • Z261874 - Combination cylinder wrench
EMAIL THIS PAGE TO A FRIEND
Z261874 Aldrich
Combination cylinder wrench
L 9 in.
Purchase
Properties
Related Categories Gas Equipment Product Guide, Gas Tools, Labware More...
packaging   pkg of 1 ea
L   9 in.
Description
General description
Ten-way wrench has openings from 7/16 to 11/8 in. Includes a boxed opening for B and MC acetylene valves, and a socket for commercial acetylene cylinders.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?