• USA Home
  • Z262285 - KNF Laboport® solid PTFE vacuum pump

EMAIL THIS PAGE TO A FRIEND
Z262285 Aldrich

KNF Laboport® solid PTFE vacuum pump

limit 10 L/min pumping speed, AC input 230 V

Purchase

Properties

Related Categories Analytical/Chromatography, Diaphragm Vacuum Pumps, HPLC, HPLC Accessories, KNF,
feature   CE compliant
mfr. no.   KNF
limit   ≤6 torr vacuum
  10 L/min pumping speed
L × W × H   11 in. × 5 1/2 in. × 7.2 in.
weight   16 lb (7.3 kg)

Description

General description

Quiet, high performance diaphragm vacuum pumps can be used alone or as the center of a modular laboratory vacuum system. Replaces noisy rotary-vane pumps for vacuum distillation, drying, filtration, rotary evaporation, degassing of liquids, and applications where water aspirators are used.

• Solid PTFE heads
• Molded PTFE diaphragm
• Kalrez® parts eliminate chemical attack to the pump
• Oil-free operation ensures pumped medium will stay pure
• New multi-port valve system with Kalrez disks improves flow and reliability
• Two stages
• 10 mm I.D. hose barbs on ports

Legal Information

Kalrez is a registered trademark of E. I. du Pont de Nemours and Company

Laboport is a registered trademark of KNF Neuberger GmbH

Price and Availability


Discover the Milli-Q® IQ 7000 ultrapure water system

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

SPE Cartridges (Tubes) - Phase Selection Guide

The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?