• USA Home
  • Z262374 - KNF Laboport® vacuum system components
EMAIL THIS PAGE TO A FRIEND
Z262374 Aldrich
KNF Laboport® vacuum system components
Inlet/exhaust separator
Purchase
Properties
Related Categories Diaphragm Vacuum Pumps, KNF, Laboport vacuum system components, Labware, Manufacturer Browser,
feature   CE compliant
mfr. no.   KNF
Description
General description
Implosion protected glass container that is used on either the inlet or exhaust side of the vacuum system. Prevents particulate from entering the pump and ensures full performance over a long period of time. Includes metal mounting cup.
Modular equipment with plug-in connectors for building systems that will support many laboratory vacuum requirements. Components do not include 3/8 in. I.D. vacuum tubing (see the Tubing Section) or tool kit, available separately. All components are sold separately.
Legal Information
Laboport is a registered trademark of KNF Neuberger GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?