• USA Home
  • Z262390 - KNF Laboport® vacuum system components
EMAIL THIS PAGE TO A FRIEND
Z262390 Aldrich
KNF Laboport® vacuum system components
Vacuum controller A, AC/DC input 115 V, 60 Hz
Purchase
Properties
Related Categories KNF, Laboport vacuum system components, Labware, Manufacturer Browser, Pumps,
feature   CE compliant
  CSA approved
mfr. no.   KNF
availability   not available in EU
  not available in EU
Description
General description
Modular equipment with plug-in connectors for building systems that will support many laboratory vacuum requirements. Components do not include 3/8 in. I.D. vacuum tubing (see the Tubing Section) or tool kit, available separately. All components are sold separately.
Vacuum controllers A solid-state module maintains precise control (±2%) of the system vacuum by cycling the pump on or off, as required. Adjustable for any level between <2 mbar and 999 mbar. Controller A includes an electronics box for connecting system components. A second vacuum controller B is required to run two processes simultaneously using the same pump.
Legal Information
Laboport is a registered trademark of KNF Neuberger GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?