• USA Home
  • Z262420 - KNF Laboport® vacuum system components
EMAIL THIS PAGE TO A FRIEND
Z262420 Aldrich
KNF Laboport® vacuum system components
Vacuum controller B, AC input 230 V50 Hz
Purchase
Properties
Related Categories Diaphragm Vacuum Pumps, KNF, Laboport vacuum system components, Labware, Manufacturer Browser,
feature   CE compliant
mfr. no.   KNF
Description
General description
A solid-state module maintains precise control (±2%) of the system vacuum by cycling the pump on or off, as required. Adjustable for any level between <2 mbar and 999 mbar. Controller A includes an electronics box for connecting system components. A second vacuum controller B is required to run two processes simultaneously using the same pump.
Modular equipment with plug-in connectors for building systems that will support many laboratory vacuum requirements. Components do not include 3/8 in. I.D. vacuum tubing (see the Tubing Section) or tool kit, available separately. All components are sold separately.
Legal Information
Laboport is a registered trademark of KNF Neuberger GmbH
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?