EMAIL THIS PAGE TO A FRIEND
Z262471 Aldrich
Chem3D Ultra
standard
Purchase
Properties
Related Categories Chemistry software, Computer Software, Labware More...
mfr. no.   Cambridgesoft
Description
Compatibility
Minimum system requirements: Windows: 95, 98, 2000, NT 4.0, Pentium, PowerPC, 32MB
General description
"Ultimate modeling, visualization and analysis"
Brings workstation quality molecular visualization and display to your desktop. Includes CS MOPAC and set-up/control interfaces for optional use of GAMESS and Gaussian. Compute advanced physical properties with CLogP and ChemProp, and create SAR tables using property servers to generate data for lists of compounds. Use ChemSAR for Excel to explore structure activity relationships and use optional Conformer for con-formational searching. Publish and view models on the web using the Chem3D Plugin. Includes ChemDraw Standard.
• Model types: space filling CPK, ball and stick, stick, ribbons, VDW dot surfaces and wire-frame
• Compute and visualize partial charges
• 3D surface properties including MEP and orbital mapping
• Create 3D models from ChemDraw or ISIS Draw
• Polypeptide builder with residue recognition
• Supports: PDB, MDL Molfile, Beilstein ROSDAL, Tripos SYBYL MOL, EPS, PICT, GIF, 3DMF, TIFF, PNG and more
• ChemProp—advanced property predictions including CLogP, BP, MP, and more
• ChemSAR for Excel builds SAR tables
• Accept output from other 3D graphics modeling packages
Legal Information
Chem3D is a trademark of CambridgeSoft
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?