• USA Home
  • Z262951 - Wheaton narrow mouth bottles with caps

EMAIL THIS PAGE TO A FRIEND
Z262951 Aldrich

Wheaton narrow mouth bottles with caps

capacity 8 oz (250 mL), clear, white polypropylene cap, vinyl flat liner

Synonym: Wheaton bottle, Wheaton bottles, narrow mouth

Purchase

Properties

material   vinyl flat liner
  white polypropylene cap
cap size   24-400
mfr. no.   Wheaton Scientific
capacity   8 oz (250 mL)
color   clear

Description

General description

Convenience packed with closures attached. Bottles made of Wheaton "900" amber glass or "800" clear soda lime flint glass.

Choice of cap styles:
• White PP cap with PTFE or polyvinyl liner
• Black plastic cap with Poly-Seal® PE cone liner

Legal Information

Poly-Seal is a registered trademark of Wheaton Industries, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?