• USA Home
  • Z263079 - Wheaton wide-mouth bottles with caps

EMAIL THIS PAGE TO A FRIEND
Z263079 Aldrich

Wheaton wide-mouth bottles with caps

capacity 8 oz (250 mL), clear, white polypropylene cap, foamed PE flat liner

Synonym: Wheaton bottle, Wheaton bottles, wide mouth glass

Purchase

Properties

material   foamed PE flat liner
  white polypropylene cap
cap size   58-400
packaging   pkg of 24 ea
mfr. no.   Wheaton Scientific
capacity   8 oz (250 mL)
color   clear

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?