• USA Home
  • Z263109 - Wheaton wide-mouth bottles with caps

EMAIL THIS PAGE TO A FRIEND
Z263109 Aldrich

Wheaton wide-mouth bottles with caps

capacity 65 oz (2000 mL), clear, white polypropylene cap, foamed PE flat liner

Synonym: Wheaton bottle, Wheaton bottles, wide mouth glass

Purchase

Properties

Related Categories Alcan Packaging, Alcan Packaging Glass Bottles, Alcan Packaging Wheaton Wide-Mouth Glass Bottles with Caps, Bottles and Caps, Glass Bottles,
material   foamed PE flat liner
  white polypropylene cap
cap size   83-400
mfr. no.   Wheaton Scientific 217401
packaging   pkg of 6 ea
capacity   65 oz (2000 mL)
diam. × H   122 mm × 213 mm
color   clear

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?