• USA Home
  • Z263303 - Wheaton straight-mouth bottles with cap, clear

EMAIL THIS PAGE TO A FRIEND
Z263303 Aldrich

Wheaton straight-mouth bottles with cap, clear

capacity 4 oz (125 mL), white polypropylene cap, foamed PE flat liner

Synonym: Wheaton bottles, bottles clear, clear bottles

Purchase

Properties

Related Categories Alcan Packaging, Alcan Packaging Glass Bottles, Alcan Packaging Wheaton Straight-Mouth Clear Glass Bottles with Cap, Bottles and Caps, Glass Bottles,
material   foamed PE flat liner
  white polypropylene cap
cap size   58-400
packaging   pkg of 24 ea
mfr. no.   Wheaton Scientific
capacity   4 oz (125 mL)

Description

Application

Ideal for soil sampling and environmental applications.

General description

Walls allow for complete removal of contents.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?