• USA Home
  • Z263559 - Adapter for Kilo-Lab® and Pure-Pac® cylinders
EMAIL THIS PAGE TO A FRIEND
Z263559 Aldrich
Adapter for Kilo-Lab® and Pure-Pac® cylinders
ferrule size 1/2 in., NPTF 1/2 in.
Purchase
Properties
Related Categories Air-Sensitive Chemistry, Aldrich Air Sensitive, Aldrich Air Sensitive Kilo-Lab, Aldrich Air Sensitive Kilo-Lab Transfer Line Adapters, Labware More...
material   stainless steel
description   with double ferrules and nuts
NPTF   1/2 in.
ferrule size   1/2 in.
Description
Legal Information
Kilo-Lab is a registered trademark of Sigma-Aldrich Co. LLC
Pure-Pac is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?