• USA Home
  • Z264423 - Orion® 370 PerpHecT® benchtop pH/ORP/ISE/T meter
EMAIL THIS PAGE TO A FRIEND
Z264423 Aldrich
Orion® 370 PerpHecT® benchtop pH/ORP/ISE/T meter
Thermo Electron Orion®, 370, AC input 115 V
Purchase
Properties
Related Categories Benchtop Meters, Labware, pH Meters, pH Supplies
feature   calibration points 3
  outputs RS232, 2.5 mm pin recorder
mfr. no.   Thermo Electron Orion®, 370
measuring range   0.00-14.00 pH
  0.000-19900 ISE conc., accuracy: 0.2 mV
  ±1000 log ORP
  ±1999.9 mV ORP, accuracy: 0.2, resolution: 0.1 mV
parameter   0-100 °C temp. range
L × W × H   9 in. × 7 in. × 2 in.
Description
General description
Orion 370 PerpHecT digital LogR meters include all the pH bascis plus simultaneous display of pH, mV or RmV (ORP) and temperature. Use autocalibration with built-in Auto-Buffer-Recognition of three buffers or enter custom buffer values. Temperature compensation can be LogR, automatic (with a combination probe) or manual. These enhanced PerHecT meters come complete with GLPcheck for verification of electrode reliability and GLPdoc fir communications. The Orion 370 meter features a concentration mode for ISE measurements which allows a two point calibration and direct measurement. LED display.
Includes meter and Orion combination Ag/AgCI electrode 9202BN and manual.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
PerpHecT is a registered trademark of Analytical Technology, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?