• USA Home
  • Z264482 - Orion® 320 & 330 PerpHecT® benchtop pH/ORP/T meter
EMAIL THIS PAGE TO A FRIEND
Z264482 Aldrich
Orion® 320 & 330 PerpHecT® benchtop pH/ORP/T meter
Thermo Electron Orion®, 320, AC input 220 V AC
Purchase
Properties
Related Categories Benchtop Meters, Labware, pH Meters, pH Supplies
feature   CE compliant
  CSA approved
  UL listed
  calibration points 2
  outputs: no
mfr. no.   Thermo Electron Orion®, 320
measuring range   0.00-14.00 pH, accuracy: 0.02, resolution: 0.01 pH unit
  80-120% pH
  ±1000 log ORP, accuracy: 1, resolution: 1 mV
  ±1999.9 ORP, accuracy: 1, resolution: 1 mV
parameter   0-100 °C temp. range
L × W × H   9 in. × 7 in. × 2 in.
Description
General description
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
PerpHecT is a registered trademark of Analytical Technology, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?