• USA Home
  • Z264547 - Orion® ion selective pH meter
EMAIL THIS PAGE TO A FRIEND
Z264547 Aldrich
Orion® ion selective pH meter
Thermo Electron Orion®, 720A, AC input 220 V
Purchase
Properties
mfr. no.   Thermo Electron Orion®, 720A
measuring range   −1,600.0-1,600.0 mV voltage, accuracy: 0.2, resolution: 0.1
  −2.000-19.999 pH, accuracy: 0.002, resolution: 0.001
  0.000-19,900 ISE, accuracy: 0.5%
L × W × H   8 in. × 9 in. × 2 in.
Description
General description
With dual electrode inputs, the Orion 720A+ allows two measurements at the same time, perfect for a busy, multi-user laboratory. The pH autocalibration feature recognizes five pH buffers using built-in buffer/temperature tables. Express concentration results to three significant digits. Full five point concentration calibration with automatic blank correction improves results of low level measurements. User prompting walks you through each meter operation. Store data using the internal datalogger or print, via the RS232 port.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?