• USA Home
  • Z264563 - Orion® ion selective pH meter
EMAIL THIS PAGE TO A FRIEND
Z264563 Aldrich
Orion® ion selective pH meter
Thermo Electron Orion®, 710A+, AC input 220 V
Purchase
Properties
mfr. no.   Thermo Electron Orion®, 710A+
measuring range   −1,600.0-1,600.0 mV voltage, accuracy: 0.2, resolution: 0.1
  −2.000-19.999 pH, accuracy: 0.005, resolution: 0.001
  0.000-19,900 ISE, accuracy: 0.5%
L × W × H   8 in. × 9 in. × 2 in.
Description
General description
This model is loaded with features to make routine pH and concentration measurements fast, easy and accurate. Meter accepts up to five standards for concentration calibration to ensure low-level measurement accuracy. Calibrate quickly using pH autocalibration with built-in buffer/temperature tables. Autoblank correction feature automatically corrects concentration results for background ion levels.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?