• USA Home
  • Z264598 - Chloride combination ISE Cl- electrode
EMAIL THIS PAGE TO A FRIEND
Z264598 Aldrich
Chloride combination ISE Cl- electrode
Purchase
Properties
Related Categories Electrodes, Labware, Manufacturer Browser, Thermo, Thermo pH,
feature   interferences OH-, S-2, Br-, CN-, NH3, S2O3-
mfr. no.   Orion
  Thermo Electron
measuring range   1.8-35,500 ppm ISE
Description
Other Notes
Suitable for use as references for ASTM D512-89
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?