• USA Home
  • Z264601 - Sodium Na+ electrode
EMAIL THIS PAGE TO A FRIEND
Z264601 Aldrich
Sodium Na+ electrode
Purchase
Properties
Related Categories Electrodes, Labware, Manufacturer Browser, Orion, Sodium Na+ electrode,
feature   interferences [1A]+, NH4+
mfr. no.   Orion
  Thermo Electron
measuring range   0.02 ppm to saturation ISE
Description
Other Notes
Suitable for use as references for ASTM D2791, AOAC 76.25
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?