• USA Home
  • Z264652 - Orion® ISE electrodes
EMAIL THIS PAGE TO A FRIEND
Z264652 Aldrich
Orion® ISE electrodes
Ammonia NH3/NH4+ electrode
Purchase
Properties
Related Categories Electrodes, ISE electrodes, Labware, Manufacturer Browser, Orion,
feature   interferences volatile amines
mfr. no.   Orion
  Thermo Electron
measuring range   0.01-14,000 ppm N ISE
Description
Other Notes
Suitable for use as references for ASTM D1426-89, EPA 350.3
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?