• USA Home
  • Z264733 - Orion® ion selective pH meter
EMAIL THIS PAGE TO A FRIEND
Z264733 Aldrich
Orion® ion selective pH meter
Thermo Electron Orion®, 920A, AC input 115 V
Purchase
Properties
mfr. no.   Thermo Electron Orion®, 920A
measuring range   −1,600.0-1,600.0 mV voltage, accuracy: 0.2, resolution: 0.1
  −2.000-19.999 pH, accuracy: 0.002, resolution: 0.001
  0.000-19,900 ISE, accuracy: 0.5%
L × W × H   10 in. × 13 in. × 4 in.
Description
General description
The Orion 920A+ provides more features than ever before, making this meter easy to use and smart. This meter accepts up to five standards for concentration calibration to ensure low-level measurement accuracy. Autoblank correction feature automatically corrects concentration results for background ion levels. Other features adding to the ease of use include high and low alarm limits and display hold. Orion 920A advanced ISE features include pre-programmed incremental techniques, calibration edit, and temperature-compensated ISE measurement. Use calibration edit feature for reviewing calibration standards to determine working range.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Orion is a registered trademark of Thermo Electron Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?