• USA Home
  • Z264903 - Hydrion® pH Jumbo dispenser
EMAIL THIS PAGE TO A FRIEND
Z264903 Aldrich
Hydrion® pH Jumbo dispenser
1-6 pH
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Hydrion, Labware,
feature   subdivision 1 pH unit
mfr. no.   Hydrion
measuring range   1-6 pH
W × L   1/2 in. × 50 ft
Description
General description
Jumbo size paper is easier to read with large size color charts. Yields a sharp color change for each pH unit.
Legal Information
Hydrion is a registered trademark of Micro Essential Laboratory, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?