• USA Home
  • Z265020 - Organomagnesium Methods In Organic Synthesis
EMAIL THIS PAGE TO A FRIEND
Z265020 Aldrich
Organomagnesium Methods In Organic Synthesis
  •  ISBN-10 0127309454
  •  ISBN-13 978-012730945-3
Purchase
Properties
Related Categories Books and Software, Inorganic and Organometallic Books, Labware More...
publication info   B. Wakefield, Academic Press, 1995, 249 pp., hard cover
mfr. no.   Elsevier
Description
General description
The book opens with a general overview of the constitution and reactivity of organomagnesium compounds, followed by information on handling them and on their detection and estimation. Throughout, practical aspects as well as principles are emphasized. The chapters on the synthesis of organomagnesium compounds cover the preparation of special forms of metallic magnesium and the reaction of magnesium with substrates such as dienes, as well as the traditional preparation of Grignard reagents. Preparations by metallation and metal-halogen exchanges are also included, as are newer methods such as hydromagnesiation of alkenes and alkynes. Systematic coverage is provided on synthetically useful reactions of organomagnesium compounds.
Table of Contents
Introduction and General References.
General Considerations in the Application of Organomagnesium Compounds in Organic and Organometallic Synthesis.
Preparation of Organomagnesium Compounds.
Addition of Organomagnesium Compounds to Carbon-Carbon Multiple Bonds.
Addition of Organomagnesium Compounds to Carbon-Nitrogen Multiple Bonds.
Addition of Organomagnesium Compounds to Carbonyl Groups.
Carbophilic Addition of Organomagnesium Compounds to Thiocarbonyl Groups.
Substitution at Carbon by Organomagnesium Compounds.
Carbenoid and Arynoid Reactions of Organomagnesium Compounds.
Reactions of Organomagnesium Compounds with Proton Donors.
Formation of Carbon-Nitrogen Bonds via Organomagnesium Compounds.
Formation of Carbon-Oxygen Bonds via Organomagnesium Compounds.
Formation of Carbon-Sulfur, Carbon-Selenium, and Carbon-Tellurium Bonds via Organomagnesium Compounds.
Formation of Carbon-Halogen Bonds via Organomagnesium Compounds.
Synthesis of Organoboron, Organosilicon, and Organophosphorous Compounds from Organomagnesium Compounds.
Organomagnesium Compounds in the Synthesis of Other Organometallic Compounds.
Index of Compounds and Methods.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?