• USA Home
  • Z265373 - Analtech HPTLC Uniplates: C18-silica gel matrix
EMAIL THIS PAGE TO A FRIEND
Z265373 Aldrich
Analtech HPTLC Uniplates: C18-silica gel matrix
Purchase
Properties
Related Categories Analtech, Analtech C18-Silica Gel Matrix, Analytical/Chromatography, Chromatography, Chromatography, TLC Plates,
material   C18-silica gel matrix
  glass support
description   reversed-phase for aqueous applications
feature   binder Organic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 55077
application(s)   thin layer chromatography (TLC): suitable
L × W   10 cm × 10 cm
layer thickness   150 μm
particle size   8-10 μm
Description
Legal Information
Uniplates is a trademark of Analtech, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?