• USA Home
  • Z265381 - Analtech HPTLC Uniplates: C2-silica gel matrix
EMAIL THIS PAGE TO A FRIEND
Z265381 Aldrich
Analtech HPTLC Uniplates: C2-silica gel matrix
Purchase
Properties
Related Categories Analtech, Analtech C2-Silica Gel Matrix, Analytical/Chromatography, Chromatography, Chromatography, TLC Plates,
material   glass support
description   reverse phase
feature   binder Organic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 08077
L × W   10 cm × 10 cm
layer thickness   150 μm
particle size   10 μm
Description
General description
High-performance TLC plates are prepared with high efficiency silica gel. The smooth homogeneous surface of the plates yields a high signal-to-noise ratio, thus increasing sensitivity and precision.
Legal Information
Uniplates is a trademark of Analtech, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?