• USA Home
  • Z265527 - Analtech TLC Uniplates: silica gel matrix
EMAIL THIS PAGE TO A FRIEND
Z265527 Aldrich
Analtech TLC Uniplates: silica gel matrix
L × W 10 cm × 20 cm, binder, Organic, fluorescent indicator
  •  eCl@ss 32110504
Purchase
Properties
Related Categories Analtech, Analtech Silica Gel Matrix, Analtech TLC Plates, Analtech TLC Plates, Uniplates, Analtech TLC Uniplates, Silica matrix,
material   glass support
  silica gel matrix
feature   binder Organic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 47021
application(s)   thin layer chromatography (TLC): suitable
L × W   10 cm × 20 cm
layer thickness   250 μm
Description
General description
Most rugged silica gel plates available.
Legal Information
Uniplates is a trademark of Analtech, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?