• USA Home
  • Z265667 - Analtech TLC Uniplates: alumina matrix
EMAIL THIS PAGE TO A FRIEND
Z265667 Aldrich
Analtech TLC Uniplates: alumina matrix
L × W 5 cm × 20 cm
  •  eCl@ss 32110504
Purchase
Properties
Related Categories Analtech, Analtech Alumina Matrix, Analtech TLC Plates, Uniplates, Analtech TLC Uniplates, Alumina matrix, Analytical/Chromatography,
material   alumina matrix
  glass support
feature   binder Inorganic
  fluorescent indicator
packaging   pkg of 25 ea
mfr. no.   Analtech, 04031
application(s)   thin layer chromatography (TLC): suitable
L × W   5 cm × 20 cm
layer thickness   250 μm
Description
General description
Produced using a slurry containing calcium sulfate hemihydrate as a binder.
Legal Information
Uniplates is a trademark of Analtech, Inc.
Price and Availability
Pharma QC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?