• USA Home
  • Z266272 - Polymer-body combination pH electrodes
EMAIL THIS PAGE TO A FRIEND
Z266272 Aldrich
Polymer-body combination pH electrodes
Standard pin fitting
Purchase
Description
General description
Long, narrow/tubes, vials. Sealed, gel-filled
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?