• USA Home
  • Z266434 - Barnant speed controller
EMAIL THIS PAGE TO A FRIEND
Z266434 Aldrich
Barnant speed controller
AC input 115 V, 50 - 60 Hz
Purchase
Properties
Related Categories Labware, Overhead - SS Stirrers, Stirrers More...
mfr. no.   Barnant
weight   4 lb (1.8 kg)
Description
General description
Solid-state speed control with a 100:1 adjustment range. Motor speed is maintained within 10% despite changes in the power or torque load conditions. Separate forward/off/reverse switch, speed dial, and LED power indicator. Controller is connected to mixer head via 6 ft power cord.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?